Invitrogen
Manufacturer Code:MA514831
Catalog # PIMA514831
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | E.425.6 |
Immunogen | An LKB1 partial fusion protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: Liver kinase B1; Liver kinase B1 homolog; LKB1; polarization-related protein LKB1; polarization-related protein LKB1 renal carcinoma antigen NY-REN-19 serine/threonine-protein kinase 11 serine/threonine-protein kinase LKB1; Renal carcinoma antigen NY-REN-19; serine/threonine-protein kinase 11; serine/threonine-protein kinase LKB1; Serine/threonine-protein kinase STK11
Gene Aliases: AA408040; hLKB1; LKB1; Par-4; PJS; R75140; STK11
UniProt ID: (Human) Q15831, (Rat) D4AE59, (Mouse) Q9WTK7
Entrez Gene ID: (Human) 6794, (Rat) 314621, (Mouse) 20869
Molecular Function: non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.