Proteintech
Manufacturer Code:167971AP150UL
Catalog # 50-173-1185
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | LITAF Fusion Protein Ag10269 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Eet-1; Estrogen-enhanced transcript protein; Estrogen-enhanced transcript protein 1; estrogen-responsive uterine transcript; lipopolysaccharide-induced TNF factor; lipopolysaccharide-induced TNF-alpha factor; Lipopolysaccharide-induced tumor necrosis factor-alpha factor; Lipopolysaccharide-induced tumor necrosis factor-alpha factor homolog; LITAF LPS induced TNF alpha factor p53 induced gene 7 protein PIG7 SIMPLE TP53I7; LITAF-like protein; LPS-induced TN factor; LPS-induced TNF-alpha factor; LPS-induced TNF-alpha factor homolog; mEET; NEDD4 WW domain-binding protein 3; p53-induced gene 7 protein; Small integral membrane protein of lysosome/late endosome; tumor protein p53 inducible protein 7
Gene Aliases: 3222402J11Rik; C85531; EET-1; LITAF; N4WBP3; PIG7; SIMPLE; TBX1; TP53I7
UniProt ID: (Human) Q99732, (Rat) P0C0T0, (Mouse) Q9JLJ0
Entrez Gene ID: (Human) 9516, (Rat) 65161, (Mouse) 56722
Molecular Function:
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.