Invitrogen
Manufacturer Code:MA519522
Catalog # PIMA519522
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 5A5 |
Immunogen | PAFAH1B1 (NP&lowbar000421 1 a.a. &bsim 110 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: LIS-1; LIS1 LIS2 MDCR MDS PAFAH LIS-1 PAF acetylhydrolase 45 kDa subunit PAF-AH 45 kDa subunit PAF-AH alpha PAFAH alpha lissencephaly 1 protein lissencephaly-1 protein platelet-activating factor acetylhydrolase IB subunit alpha platelet-activating factor acetylhydrolase isoform Ib alpha subunit (45kD) platelet-activating factor acetylhydrolase isoform Ib subunit 1 (45kDa); lissencephaly 1 protein; Lissencephaly-1 protein; PAF acetylhydrolase 45 kDa subunit; PAF-AH 45 kDa subunit; PAF-AH alpha; PAFAH alpha; PAFAH1B1; platelet-activating factor acetylhydrolase 1b, regulatory subunit 1 (45kDa); Platelet-activating factor acetylhydrolase IB subunit beta; platelet-activating factor acetylhydrolase, isoform Ib, alpha subunit (45kD); platelet-activating factor acetylhydrolase, isoform Ib, subunit 1 (45kDa)
Gene Aliases: LIS1; LIS2; MDCR; MDS; NudF; PAFAH; PAFAH1B1; PAFAHA
UniProt ID: (Human) P43034
Entrez Gene ID: (Human) 5048
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.