Invitrogen
Manufacturer Code:PA582710
Catalog # PIPA582710
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human PAFAH1B1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: LIS-1; LIS1 MDCR MDS PAFAH; lissencephaly 1 protein; Lissencephaly-1 protein; PAF acetylhydrolase 45 kDa subunit; PAF-AH 45 kDa subunit; PAF-AH alpha; PAFAH alpha; PAFAH1B1; platelet-activating factor acetylhydrolase 1b, regulatory subunit 1 (45kDa); Platelet-activating factor acetylhydrolase IB subunit beta; platelet-activating factor acetylhydrolase, isoform Ib, alpha subunit (45kD); platelet-activating factor acetylhydrolase, isoform Ib, subunit 1 (45kDa)
Gene Aliases: LIS1; LIS2; MDCR; MDS; NudF; PAFAH; PAFAH1B1; PAFAHA
UniProt ID: (Human) P43034
Entrez Gene ID: (Human) 5048
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.