Invitrogen
Manufacturer Code:PA570188
Catalog # PIPA570188
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the N terminal region of human LIS1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: LIS-1; LIS-1 {ECO:0000255|HAMAP-Rule:MF_03141}; LIS1 LIS2 MDCR MDS PAFAH LIS-1 PAF acetylhydrolase 45 kDa subunit PAF-AH 45 kDa subunit PAF-AH alpha PAFAH alpha lissencephaly 1 protein lissencephaly-1 protein platelet-activating factor acetylhydrolase IB subunit alpha platelet-activating factor acetylhydrolase isoform Ib alpha subunit (45kD) platelet-activating factor acetylhydrolase isoform Ib subunit 1 (45kDa); lissencephaly 1 protein; Lissencephaly-1 protein; lissencephaly-1 protein {ECO:0000255|HAMAP-Rule:MF_03141}; Miller-Dieker syndrome chromosome region; PAF acetylhydrolase 45 kDa subunit; PAF acetylhydrolase 45 kDa subunit {ECO:0000255|HAMAP-Rule:MF_03141}; PAF-AH 45 kDa subunit; PAF-AH 45 kDa subunit {ECO:0000255|HAMAP-Rule:MF_03141}; PAF-AH alpha; PAF-AH alpha {ECO:0000255|HAMAP-Rule:MF_03141}; PAF-AH beta; PAFAH alpha; PAFAH alpha {ECO:0000255|HAMAP-Rule:MF_03141}; PAFAH1B1; platelet-activating factor acetylhydrolase 1b, regulatory subunit 1; platelet-activating factor acetylhydrolase 1b, regulatory subunit 1 (45kDa); platelet-activating factor acetylhydrolase beta subunit (PAF-AH beta); Platelet-activating factor acetylhydrolase IB subunit alpha; platelet-activating factor acetylhydrolase IB subunit alpha {ECO:0000255|HAMAP-Rule:MF_03141}; Platelet-activating factor acetylhydrolase IB subunit beta; platelet-activating factor acetylhydrolase, isoform 1b, beta1 subunit; platelet-activating factor acetylhydrolase, isoform 1b, subunit 1; platelet-activating factor acetylhydrolase, isoform Ib, alpha subunit (45kD); platelet-activating factor acetylhydrolase, isoform Ib, alpha subunit 45kDa; platelet-activating factor acetylhydrolase, isoform Ib, subunit 1 (45kDa)
Gene Aliases: LIS-1; LIS1; LIS2; MDCR; MDS; Mdsh; MMS10-U; Ms10u; NudF; PAFAH; PAFAH1B1; PAFAHA
UniProt ID: (Bovine) P43033, (Human) P43034, (Rat) P63004, (Mouse) P63005
Entrez Gene ID: (Horse) 100072490, (Bovine) 282513, (Rabbit) 100354130, (Human) 5048, (Dog) 480656, (Rat) 83572, (Guinea pig) 100734716, (Mouse) 18472
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.