Novus Biologicals
Manufacturer Code:NB1001040
Catalog # NB1001040
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence TGSVDQTVKVWECR corresponding to C-Terminus according to NP_000421.1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: LIS-1; LIS-1 LIS1PAF-AH alpha LIS2 lissencephaly 1 protein Lissencephaly-1 protein MDCR MDS Miller-Dieker syndrome chromosome region PAF acetylhydrolase 45 kDa subunit PAFAH PAF-AH 45 kDa subunit PAFAH alpha PAFAHA platelet-activating factor acetylhydrolase 1b regulatory subunit 1 (45kDa) platelet-activating factor acetylhydrolase IB subunit alpha platelet-activating factor acetylhydrolase isoform Ib alpha subunit (45kD) platelet-activating factor acetylhydrolase isoform Ib alpha subunit 45kDa platelet-activating factor acetylhydrolase isoform Ib subunit 1 (45kDa); lissencephaly 1 protein; Lissencephaly-1 protein; PAF acetylhydrolase 45 kDa subunit; PAF-AH 45 kDa subunit; PAF-AH alpha; PAFAH alpha; PAFAH1B1; platelet-activating factor acetylhydrolase 1b, regulatory subunit 1 (45kDa); Platelet-activating factor acetylhydrolase IB subunit beta; platelet-activating factor acetylhydrolase, isoform Ib, alpha subunit (45kD); platelet-activating factor acetylhydrolase, isoform Ib, subunit 1 (45kDa)
Gene Aliases: LIS1; LIS2; MDCR; MDS; NudF; PAFAH; PAFAH1B1; PAFAHA
UniProt ID: (Human) P43034
Entrez Gene ID: (Human) 5048
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.