Invitrogen
Manufacturer Code:PA559119
Catalog # PIPA559119
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human LIPT2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Lipoate-protein ligase B; Lipoyl/octanoyl transferase; Octanoyl-[acyl-carrier-protein]-protein N-octanoyltransferase; Putative lipoyltransferase 2 mitochondrial Lipoate-protein ligase B Lipoyl/octanoyl transferase Octanoyl-&lsqbacyl-carrier-protein]-protein N-octanoyltransferase LIPT2; Putative lipoyltransferase 2, mitochondrial; putative octanoyltransferase, mitochondrial
Gene Aliases: LIPT2
UniProt ID: (Human) A6NK58
Entrez Gene ID: (Human) 387787
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.