Invitrogen
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | 1:1,000 |
Host / Isotype | Rabbit / IgG |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 13-43 amino acids from the N-terminal region of human LIPT2. |
Conjugate | Unconjugated |
Form | Liquid |
Concentration | Lot-Specific |
Storage conditions | -20° C, Avoid Freeze/Thaw Cycles |
RRID | AB_2634428 |
Catalyzes the transfer of endogenously produced octanoic acid from octanoyl-acyl-carrier-protein onto the lipoyl domains of lipoate-dependent enzymes. Lipoyl-ACP can also act as a substrate although octanoyl-ACP is likely to be the physiological substrate.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.