Novus Biologicals
Manufacturer Code:NBP2831450.1ML
Catalog # NB031074
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the C-terminal region of Human LIPT2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Lipoate-protein ligase B; lipoyl(octanoyl) transferase 2 (putative) mitochondrial octanoyl-acyl-carrier-protein-protein N-octanoyltransferase putative octanoyltransferase mitochondrial; Lipoyl/octanoyl transferase; Octanoyl-[acyl-carrier-protein]-protein N-octanoyltransferase; Putative lipoyltransferase 2, mitochondrial; putative octanoyltransferase, mitochondrial
Gene Aliases: LIPT2
UniProt ID: (Human) A6NK58
Entrez Gene ID: (Human) 387787
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.