Invitrogen
Manufacturer Code:PA543148
Catalog # PIPA543148
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human LIPF |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: gastric lipase; Gastric triacylglycerol lipase; GL; Lingual lipase; lipase F; Lipase gastric GL HGL HLAL MGC138477 MGC142271; lipase, gastric; prelingual lipase; triacylglycerol lipase
Gene Aliases: 2310051B21Rik; AV082900; DGL; GL; HGL; HLAL; LIPF; RNLIP; Rnlp
UniProt ID: (Human) P07098, (Dog) P80035, (Rat) P04634, (Mouse) Q9CPP7
Entrez Gene ID: (Human) 8513, (Dog) 403867, (Rabbit) 100338083, (Guinea pig) 100724215, (Rat) 50682, (Mouse) 67717
Molecular Function: hydrolase lipase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.