Invitrogen
Manufacturer Code:PA5103097
Catalog # PIPA5103097
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human LIPF corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2310051B21Rik AV082900 EDL EL endothelial lipase gastric lipase gastric triacylglycerol lipase GL HGL HLAL Lingual lipase lipase F lipase F gastric type lipase gastric LIPE LIPF MGC138477 MGC142271 prelingual lipase RNLIP Rnlp; gastric lipase; Gastric triacylglycerol lipase; GL; lipase, gastric
Gene Aliases: 2310051B21Rik; AV082900; GL; HGL; HLAL; LIPF
UniProt ID: (Human) P07098, (Mouse) Q9CPP7
Entrez Gene ID: (Human) 8513, (Mouse) 67717
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.