Invitrogen
Manufacturer Code:PA537375
Catalog # PIPA537375
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length human LIPC |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hepatic lipase; Hepatic triacylglycerol lipase; Hepatic triacylglycerol lipase Short name&equalsHL Hepatic lipase Lipase member C LIPC HTGL; HL; Lipase member C; lipase, hepatic; Lysophospholipase; Phospholipase A1; Triacylglycerol lipase
Gene Aliases: HDLCQ12; HL; HTGL; LIPC; LIPH
UniProt ID: (Human) P11150
Entrez Gene ID: (Human) 3990
Molecular Function:
esterase
hydrolase
lipase
storage protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.