Invitrogen
Manufacturer Code:PA536962
Catalog # PIPA536962
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to the 349-394 of human LIPC |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hepatic lipase; Hepatic triacylglycerol lipase; Hepatic triacylglycerol lipase Short name&equalsHL Hepatic lipase Lipase member C LIPC HTGL; HL; lipase C; Lipase member C; lipase, hepatic; Lysophospholipase; Phospholipase A1; Triacylglycerol lipase
Gene Aliases: AI256194; HDLCQ12; HL; Hpl; HTGL; LIPC; LIPH
UniProt ID: (Human) P11150, (Mouse) P27656
Entrez Gene ID: (Human) 3990, (Rat) 24538, (Mouse) 15450
Molecular Function:
esterase
hydrolase
lipase
storage protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.