Novus Biologicals
Manufacturer Code:NBP30492220UL
Catalog # NB094529
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 280-499 of human LIPC (NP_000227.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 3.1.1 EC 3.1.1.3 Hepatic lipase hepatic triacylglycerol lipase HLHDLCQ12 HTGL Lipase member C lipase hepatic LIPH; hepatic lipase; Hepatic triacylglycerol lipase; HL; Lipase member C; lipase, hepatic; Lysophospholipase; Phospholipase A1; Triacylglycerol lipase
Gene Aliases: HDLCQ12; HL; HTGL; LIPC; LIPH
UniProt ID: (Human) P11150
Entrez Gene ID: (Human) 3990
Molecular Function:
esterase
hydrolase
lipase
storage protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.