Invitrogen
Manufacturer Code:MA515278
Catalog # PIMA515278
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 7G6D7,9G7F12 |
Immunogen | Purified recombit fragment of LAL expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Acid cholesteryl ester hydrolase; cholesterol ester hydrolase; Cholesteryl esterase; LAL CESD LIPA; Lipase A; lipase A, lysosomal acid, cholesterol esterase; lysosomal acid lipase; Lysosomal acid lipase/cholesteryl ester hydrolase; Sterol esterase
Gene Aliases: CESD; LAL; LIPA
UniProt ID: (Human) P38571
Entrez Gene ID: (Human) 3988
Molecular Function: hydrolase lipase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.