Invitrogen
Manufacturer Code:PA523080
Catalog # PIPA523080
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide made to an internal region of the human Lysosomal acid lipase protein (within residues 150-300). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acid cholesteryl ester hydrolase; cholesterol ester hydrolase; Cholesteryl esterase; Lipase A; lipase A, lysosomal acid, cholesterol esterase; lysosomal acid lipase; lysosomal acid lipase 1; Lysosomal acid lipase/cholesteryl ester hydrolase; RP11-341B24.1 CESD LAL acid cholesteryl ester hydrolase cholesterol ester hydrolase cholesteryl esterase lysosomal acid lipase lysosomal acid lipase/cholesteryl ester hydrolase sterol esterase; Sterol esterase
Gene Aliases: AA960673; CESD; LAL; Lip-1; Lip1; LIPA
UniProt ID: (Human) P38571, (Mouse) Q9Z0M5
Entrez Gene ID: (Human) 3988, (Mouse) 16889
Molecular Function: hydrolase lipase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.