Invitrogen
Manufacturer Code:PA5109918
Catalog # PIPA5109918
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 110-399 of human LIPA (NP_001121077.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AA960673 Acid cholesteryl ester hydrolase CESD Chole Chole2 Cholesteryl esterase Lal Lip1 Lip-1 LIPA Lipase A Lysosomal acid lipase/cholesteryl ester hydrolase RP11-341B24.1 Sterol esterase; Acid cholesteryl ester hydrolase; cholesterol ester hydrolase; Cholesterol esterase (pancreatic) see D3Wox12 D3Wox13 D3Wox26 and D3Mgh25; Cholesteryl esterase; Lipase A; lipase A, lysosomal acid, cholesterol esterase; lysosomal acid lipase; lysosomal acid lipase 1; lysosomal acid lipase A; Lysosomal acid lipase/cholesteryl ester hydrolase; pancreatic cholesterol esterase; Sterol esterase
Gene Aliases: CESD; Chole; Chole2; LAL; Lip1; LIPA
UniProt ID: (Human) P38571
Entrez Gene ID: (Human) 3988, (Rat) 25055
Molecular Function: hydrolase lipase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.