Invitrogen
Manufacturer Code:PA542261
Catalog # PIPA542261
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human LIN7C |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: lin 7 homolog C; lin-7 homolog C; Lin-7 homolog C (C. elegans) FLJ11215 LIN-7-C LIN-7C MALS-3 VELI3 MALS3; LIN-7 protein 3; lin-7-C; Lin-7C; Lin7c; MALS-3; Mammalian lin-seven protein 3; mLin7C; neuroepithelial polarity protein; Protein lin-7 homolog C; Veli-3; veli-3 protein; vertebrate homolog of C. elegans Lin-7 type 3; Vertebrate lin-7 homolog 3
Gene Aliases: 9130007B12Rik; AI303698; AU019331; AW125731; D2Ertd520e; fb75b09; LIN-7-C; LIN-7C; LIN7C; MALS-3; MALS3; VELI3; wu:fb75b09; zgc:101881
UniProt ID: (Bovine) Q0P5F3, (Human) Q9NUP9, (Rat) Q792I0, (Mouse) O88952
Entrez Gene ID: (Dog) 485424, (Bovine) 510125, (Rabbit) 100356689, (Horse) 100072314, (Human) 55327, (Rat) 60442, (Mouse) 22343, (Guinea pig) 100729709, (Zebrafish) 447934
Molecular Function: cell adhesion molecule cell junction protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.