OriGene
Manufacturer Code:CF501435
Catalog # 50-166-0452
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1B2 |
Immunogen | Full length human recombit protein of human LIN7B produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: hLin7B; hVeli2; lin-7 homolog B; Lin-7B; LIN-7B MALS-2 MALS2 VELI2; MALS-2; Mammalian lin-seven protein 2; Protein lin-7 homolog B; Veli-2; Vertebrate lin-7 homolog 2
Gene Aliases: LIN-7B; LIN7B; MALS-2; MALS2; UNQ3116/PRO10200; VELI2
UniProt ID: (Human) Q9HAP6
Entrez Gene ID: (Human) 64130
Molecular Function: cell adhesion molecule cell junction protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.