Invitrogen
Manufacturer Code:PA581517
Catalog # PIPA581517
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to the center region of the Human LIN7A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Lin-7A; LIN-7A LIN7 MALS-1 TIP-33 VELI1 hLin-7 mammalian LIN-7 1 mammalian lin-seven protein 1 protein lin-7 homolog A tax interaction protein 33 veli-1 vertebrate LIN7 homolog 1 vertebrate lin-7 homolog 1; MALS-1; mammalian LIN-7 1; Mammalian lin-seven protein 1; Protein lin-7 homolog A; Tax interaction protein 33; TIP-33; Veli-1; Vertebrate lin-7 homolog 1; vertebrate LIN7 homolog 1
Gene Aliases: LIN-7A; LIN7; LIN7A; MALS-1; MALS1; TIP-33; VELI1
UniProt ID: (Human) O14910
Entrez Gene ID: (Human) 8825
Molecular Function: cell adhesion molecule cell junction protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.