Novus Biologicals
Manufacturer Code:NBP243587
Catalog # NBP243587
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 377 |
Immunogen | Recombinant protein encompassing a sequence within the center region of human LIN7A. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: hLin-7 LIN7 lin-7 homolog A (C. elegans) LIN-7A MALS1 MALS-1MGC148143 Mammalian lin-seven protein 1 Tax interaction protein 33 TIP-33mammalian LIN-7 1 veli-1 VELI1protein lin-7 homolog A vertebrate LIN7 homolog 1 Vertebrate lin-7 homolog 1; Lin-7A; MALS-1; mammalian LIN-7 1; Mammalian lin-seven protein 1; Protein lin-7 homolog A; Tax interaction protein 33; TIP-33; Veli-1; Vertebrate lin-7 homolog 1; vertebrate LIN7 homolog 1
Gene Aliases: LIN-7A; LIN7; LIN7A; MALS-1; MALS1; TIP-33; VELI1
UniProt ID: (Human) O14910
Entrez Gene ID: (Human) 8825
Molecular Function: cell adhesion molecule cell junction protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.