Proteintech
Manufacturer Code:550031AP150UL
Catalog # 50-173-5924
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: abnormal cell LINeage LIN-41; B-box zinc finger, Filamin and NHL repeat containing protein (123.8 kD) (lin-41); Drosophila dappled/ vertebrate TRipartite Motif protein related; E3 ubiquitin-protein ligase TRIM71; homolog of C. elegans Lin-41; LIN 41 LIN41 Protein lin 41 homolog TRIM71 tripartite motif containing 71; lin-41 homolog; mLin41; Protein lin-41 homolog; RING-type E3 ubiquitin transferase TRIM71; tripartite motif containing 71, E3 ubiquitin protein ligase; tripartite motif-containing 71; Tripartite motif-containing protein 71
Gene Aliases: 2610206G21Rik; AL022943; Gm1127; LIN-41; LIN41; mlin-41; mLin41; RGD1566388; TRIM71
UniProt ID: (Human) Q2Q1W2, (Mouse) Q1PSW8, (Rat) D3ZVM4
Entrez Gene ID: (Human) 131405, (Mouse) 636931, (Rat) 301042
Molecular Function: ubiquitin-protein ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.