Novus Biologicals
Manufacturer Code:38710002
Catalog # 38-710-002
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | In vivo generated recombinant protein fragment |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Protein Aliases: Abnormal cell lineage protein 53; Probable histone-binding protein lin-53; Protein LIN-53; similar to mammalian proteins RbAp48 and RbAp46; Synthetic multivulva protein p48
Gene Aliases: CELE_K07A1.12; K07A1.12; lin-53; rba-2
UniProt ID: (C. elegans) P90916
Entrez Gene ID: (C. elegans) 172802
Molecular Function: receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.