Invitrogen
Manufacturer Code:PA5109915
Catalog # PIPA5109915
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human LIMD1 (NP_055055.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AW822033 BOS_21185 D9Ertd192e LIM domain-containing protein 1 LIMD1; LIM domain-containing protein 1
Gene Aliases: AW822033; D9Ertd192e; LIMD1
UniProt ID: (Human) Q9UGP4, (Mouse) Q9QXD8
Entrez Gene ID: (Human) 8994, (Mouse) 29806
Molecular Function: transcription cofactor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.