Abnova
Manufacturer Code:PAB19613
Catalog # 89-121-141
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to N-terminus of human LIMD1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. Do not freeze. |
Rabbit polyclonal antibody raised against synthetic peptide of LIMD1.
For Research Use Only
Protein Aliases: LIM domain-containing protein 1
Gene Aliases: LIMD1
UniProt ID: (Human) Q9UGP4
Entrez Gene ID: (Human) 8994
Molecular Function: transcription cofactor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.