OriGene
Manufacturer Code:CF808537
Catalog # 50-166-8924
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1A3 |
Immunogen | Human recombit protein fragment corresponding to amino acids 1-275 of human LIMA1 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 1110021C24Rik; Epithelial protein lost in neoplasm; epithelial protein lost in neoplasm beta; EPLIN SREBP3; LIM domain and actin-binding protein 1; sterol regulatory element binding protein 3
Gene Aliases: EPLIN; LIMA1; PP624; SREBP3
UniProt ID: (Human) Q9UHB6
Entrez Gene ID: (Human) 51474
Molecular Function:
actin family cytoskeletal protein
cytoskeletal protein
nuclease
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.