Novus Biologicals
Manufacturer Code:NBP201926
Catalog # NBP201926
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2D5 |
Immunogen | Human recombinant protein fragment corresponding to amino acids 100-362 of human LHX1(NP_005559) produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: hLim-1; hLim-1 Homeobox protein Lim-1 LIM homeobox 1 LIM homeobox protein 1 LIM-1LIM/homeobox protein Lhx1 LIM1MGC138141 MGC126723; Homeobox protein Lim-1; LIM homeobox protein 1; LIM/homeobox protein Lhx1
Gene Aliases: LHX1; LIM-1; LIM1
UniProt ID: (Human) P48742
Entrez Gene ID: (Human) 3975
Molecular Function:
RNA binding protein
actin family cytoskeletal protein
cytoskeletal protein
helix-turn-helix transcription factor
homeobox transcription factor
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.