Invitrogen
Manufacturer Code:PA546855
Catalog # PIPA546855
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human LHX1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hLim-1; Homeobox protein Lim-1; LIM homeo box protein 1; LIM homeobox 1 LIM1 LIM-1; LIM homeobox protein 1; LIM/homeobox protein Lhx1; Rlim
Gene Aliases: LHX1; LIM-1; LIM1
UniProt ID: (Human) P48742, (Mouse) P63006, (Rat) P63007
Entrez Gene ID: (Human) 3975, (Bovine) 786041, (Mouse) 16869, (Rat) 257634
Molecular Function: RNA binding protein actin family cytoskeletal protein cytoskeletal protein helix-turn-helix transcription factor homeobox transcription factor nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.