Abnova
Manufacturer Code:MAB12924
Catalog # 89-934-481
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 4C11 |
Immunogen | Recombinant protein corresponding to full length human LIG3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at &minus20°C. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against full length recombinant human LIG3.
For Research Use Only
Protein Aliases: DNA ligase 3; DNA ligase III; LIG2; ligase II, DNA, ATP-dependent; ligase III, DNA, ATP-dependent; Polydeoxyribonucleotide synthase [ATP] 3
Gene Aliases: LIG2; LIG3
UniProt ID: (Human) P49916
Entrez Gene ID: (Human) 3980
Molecular Function:
DNA binding protein
DNA ligase
ligase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.