Novus Biologicals
Manufacturer Code:NBP211774APCCY7
Catalog # N211774APC7
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | MM0455-9B23 |
Immunogen | Human recombinant LIFRalpha extracellular domain. |
Conjugate | APC-Cyanine7 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. Do not freeze. |
Mouse Monoclonal Antibody
Protein Aliases: CD118; CD118 antigen; CD118 CD118 antigen FLJ98106 FLJ99923 leukemia inhibitory factor receptor leukemia inhibitory factor receptor alpha LIF receptor LIF-R SJS2 STWS SWS; Leukemia inhibitory factor receptor; LIF receptor
Gene Aliases: CD118; LIF-R; LIFR; SJS2; STWS; SWS
UniProt ID: (Human) P42702
Entrez Gene ID: (Human) 3977
Molecular Function: cytokine defense/immunity protein signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.