R&D Systems
Manufacturer Code:MAB5990
Catalog # MAB5990
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 673602 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse LIF R alpha Leu44-Ser828 Accession # P42703 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: CD118; CD118 antigen; CD118 CD118 antigen FLJ98106 FLJ99923 leukemia inhibitory factor receptor leukemia inhibitory factor receptor alpha LIF R alpha LIF R beta LIF receptor LIFR LIF-R LIFRa SJS2 STWS SWS; Leukemia inhibitory factor receptor; LIF receptor
Gene Aliases: CD118; LIF-R; LIFR; SJS2; STWS; SWS
UniProt ID: (Human) P42702
Entrez Gene ID: (Human) 3977
Molecular Function:
cytokine
defense/immunity protein
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.