Proteintech
Manufacturer Code:227791AP150UL
Catalog # 50-173-3942
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | LIFR Fusion Protein Ag18742 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CD118; CD118 antigen; CD118 LIF R LIF receptor LIFR SJS2 STWS SWS; D-factor/LIF receptor; Leukemia inhibitory factor receptor; leukemia inhibitory factor receptor alpha chain; LIF receptor; LIF-R; soluble differentiation-stimulating factor receptor
Gene Aliases: A230075M04Rik; AW061234; CD118; LIF; LIF-R; LIFR; SJS2; STWS; SWS
UniProt ID: (Human) P42702, (Mouse) P42703
Entrez Gene ID: (Human) 3977, (Mouse) 16880
Molecular Function:
cytokine
defense/immunity protein
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.