Novus Biologicals
Manufacturer Code:NBP176554
Catalog # NBP176554
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Blocking Assay (BLOCK) | Assay dependent |
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Neutralization (Neu) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Antibody was raised against a 16 amino acid synthetic peptide near the center of human LIF. The immunogen is located within amino acids 50 - 100 of LIF. Amino Acid Squence: LYYTAQGEPFPNNLDK |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CDF D factor DIA differentiation inhibitory activity differentiation stimulating factor Differentiation-stimulating factor Emfilermin HILDAcholinergic differentiation factor leukemia inhibitory factor leukemia inhibitory factor (cholinergic differentiation factor) Melanoma-derived LPL inhibitor MLPLI; cholinergic differentiation factor; D factor; differentiation inhibitory activity; differentiation-inducing factor; Differentiation-stimulating factor; Emfilermin; hepatocyte-stimulating factor III; human interleukin in DA cells; Leukemia inhibitory factor; LIF; Melanoma-derived LPL inhibitor; MLPLI
Gene Aliases: CDF; DIA; HILDA; LIF; MLPLI
UniProt ID: (Human) P15018
Entrez Gene ID: (Human) 3976
Molecular Function:
cytokine
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.