R&D Systems
Manufacturer Code:AB449NA
Catalog # AB449NA
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Neutralization (Neu) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant mouse LIF |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: CDF D factor DIA differentiation inhibitory activity differentiation stimulating factor Differentiation-stimulating factor Emfilermin HILDA HILDAcholinergic differentiation factor leukemia inhibitory factor leukemia inhibitory factor (cholinergi; cholinergic differentiation factor; D factor; differentiation inhibitory activity; differentiation-inducing factor; Differentiation-stimulating factor; Emfilermin; hepatocyte-stimulating factor III; human interleukin in DA cells; Leukemia inhibitory factor; LIF; Melanoma-derived LPL inhibitor; MLPLI
Gene Aliases: CDF; DIA; HILDA; LIF; MLPLI
UniProt ID: (Human) P15018
Entrez Gene ID: (Human) 3976
Molecular Function: cytokine signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.