Invitrogen
Manufacturer Code:PA522387
Catalog # PIPA522387
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein fragment contain a sequence corresponding to a region within amino acids 54 and 258 (O43766) of Lipoic acid synthetase |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HUSSY-01 LAS LIP1 LS lip-syn lipoate synthase lipoic acid synthase lipoyl synthase mitochondrial; LIAS; lip-syn; Lipoate synthase; Lipoic acid synthase; Lipoyl synthase, mitochondrial; LS
Gene Aliases: HGCLAS; HUSSY-01; LAS; LIAS; LIP1; LS; PDHLD
UniProt ID: (Human) O43766
Entrez Gene ID: (Human) 11019
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.