Novus Biologicals
Manufacturer Code:NBP15432020UL
Catalog # NBP15432020
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to LIAS(lipoic acid synthetase) The peptide sequence was selected from the C terminal of LIAS. Peptide sequence EYITPEKFKYWEKVGNELGFHYTASGPLVRSSYKAGEFFLKNLVAKRKTK. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 2.8.1.8 LASmitochondrial LIP1 lipoic acid synthetase Lip-syn MGC23245; LIAS; lip-syn; Lipoate synthase; Lipoic acid synthase; Lipoyl synthase, mitochondrial; LS
Gene Aliases: HGCLAS; HUSSY-01; LAS; LIAS; LIP1; LS; PDHLD
UniProt ID: (Human) O43766
Entrez Gene ID: (Human) 11019
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.