Proteintech
Manufacturer Code:115771AP150UL
Catalog # 50-172-7281
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | LIAS Fusion Protein Ag2162 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HUSSY 01 LAS LIAS Lip syn LIP1 Lipoate synthase Lipoic acid synthase lipoic acid synthetase Lipoyl synthase mitochondrial LS; LIAS; lip-syn; lip-syn {ECO:0000255|HAMAP-Rule:MF_03123}; Lipoate synthase; lipoate synthase {ECO:0000255|HAMAP-Rule:MF_03123}; Lipoic acid synthase; lipoic acid synthase {ECO:0000255|HAMAP-Rule:MF_03123}; Lipoyl synthase, mitochondrial; lipoyl synthase, mitochondrial {ECO:0000255|HAMAP-Rule:MF_03123}; LS; LS {ECO:0000255|HAMAP-Rule:MF_03123}
Gene Aliases: 2900022L22Rik; 4933425M12Rik; 7a5ex; C77512; HGCLAS; HUSSY-01; LAS; LIAS; LIP1; LS; mLip1; PDHLD
UniProt ID: (Human) O43766, (Rat) Q5XIH4, (Mouse) Q99M04
Entrez Gene ID: (Human) 11019, (Rat) 305348, (Mouse) 79464
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.