Invitrogen
Manufacturer Code:PA588722
Catalog # PIPA588722
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 1-70 of human LHX9 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: LIM homeobox 9-like; LIM homeobox protein 9; LIM-homeodomain type transcription factor 9; LIM/homeobox protein Lhx9
Gene Aliases: 3110009O07Rik; BB104635; LH2B; LHX9
UniProt ID: (Human) Q9NQ69, (Rat) Q80W90, (Mouse) Q9WUH2
Entrez Gene ID: (Human) 56956, (Rat) 289048, (Mouse) 16876
Molecular Function: actin family cytoskeletal protein cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.