Invitrogen
Manufacturer Code:PA541160
Catalog # PIPA541160
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human LHX9 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Lhx9; LIM homeobox 9; LIM homeobox 9-like; LIM homeobox protein 9; LIM homeodomain type transcription factor Lhx9; LIM-homeobox protein 9; LIM-homeodomain type transcription factor 9; LIM/homeobox protein Lhx9
Gene Aliases: 3110009O07Rik; BB104635; LH2B; LHX9; si:dkey-121a9.2; zgc:112196
UniProt ID: (Human) Q9NQ69, (Bovine) A0JNI8, (Rat) Q80W90, (Mouse) Q9WUH2, (Zebrafish) Q1LWV4
Entrez Gene ID: (Human) 56956, (Pig) 397597, (Dog) 490257, (Horse) 100051569, (Rabbit) 100344590, (Bovine) 515012, (Guinea pig) 100713030, (Rat) 289048, (Mouse) 16876, (Zebrafish) 550405
Molecular Function:
actin family cytoskeletal protein
cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.