Abnova
Manufacturer Code:PAB29907
Catalog # 89-033-195
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to amino acids 324-373 of human LHX9. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C for up to one week. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: LIM homeobox protein 9; LIM/homeobox protein Lhx9
Gene Aliases: LHX9
UniProt ID: (Human) Q9NQ69
Entrez Gene ID: (Human) 56956
Molecular Function: actin family cytoskeletal protein cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.