Novus Biologicals
Manufacturer Code:NBP254487AF350
Catalog # NP24487A30
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | LHCGR/1416 |
Immunogen | Purified recombinant N-terminal fragment of human LHCGR (exact sequence is proprietary) |
Conjugate | Alexa Fluor 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: FLJ41504 HHG hypergonadotropic hypogonadism LCGRLH/CGR LH/CG-R LHRHR LHRLGR2ULG5 LSH-R Luteinizing hormone receptor luteinizing hormone/choriogonadotropin receptor lutropin/choriogonadotropin receptor lutropin-choriogonadotropic hormone receptor; hypergonadotropic hypogonadism; LH/CG-R; LHR; Luteinizing hormone receptor; Lutropin-choriogonadotropic hormone receptor; lutropin/choriogonadotropin receptor
Gene Aliases: HHG; LCGR; LGR2; LH/CG-R; LH/CGR; LHCGR; LHR; LHRHR; LSH-R; ULG5
UniProt ID: (Human) P22888
Entrez Gene ID: (Human) 3973
Molecular Function:
extracellular matrix protein
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.