Invitrogen
Manufacturer Code:PA579598
Catalog # PIPA579598
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence at the N-terminus of human hCG receptoR. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hypergonadotropic hypogonadism; LH receptor; LH/CG-R; LHR; LSH-R; Luteinizing hormone receptor; Lutropin-choriogonadotropic hormone receptor; Lutropin-choriogonadotropic hormone receptor LH/CG-R Luteinizing hormone receptor LHR LSH-R LHCGR LCGR LGR2 LHRHR; lutropin/choriogonadotropin receptor
Gene Aliases: HHG; LCGR; LGR2; LH/CG-R; LH/CGR; LHCGR; LHR; LHRHR; LSH-R; LSHR; ULG5
UniProt ID: (Human) P22888, (Rat) P16235
Entrez Gene ID: (Human) 3973, (Rat) 25477
Molecular Function: extracellular matrix protein receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.