Novus Biologicals
Manufacturer Code:NBP193715
Catalog # NBP193715
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:IISFPALTFLNNHDQPFMQELVDGLYHTGANVESFSSSTRPGQMEISFLPEFGISSADNAFTLRTGVKEVARKYNYIASFFIETGFCDSGI |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GLULD1 glutamate-ammonia ligase (glutamine synthase) domain containing 1 glutamate-ammonia ligase (glutamine synthetase) domain containing 1 Glutamate-ammonia ligase domain-containing protein 1 lengsin lengsin lens protein with glutamine synthetase domain Lens glutamine synthase-like LGSMGC163240 MGC163238; glutamate-ammonia ligase (glutamine synthase) domain containing 1; Glutamate-ammonia ligase domain-containing protein 1; Lengsin; Lens glutamine synthase-like
Gene Aliases: GLULD1; LGS; LGSN
UniProt ID: (Human) Q5TDP6
Entrez Gene ID: (Human) 51557
Molecular Function: ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.