Invitrogen
Manufacturer Code:PA534171
Catalog # PIPA534171
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic 20 amino acid peptide from N-terminal extracellular domain of human RXFP1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: leucine-rich repeat-containing G protein-coupled receptor 7; Leucine-rich repeat-containing G-protein coupled receptor 7; Relaxin family peptide receptor 1; Relaxin receptor 1; relaxin/insulin-like family peptide receptor 1; RXFP1 LGR7 LGR7.1 LGR7.10 LGR7.2 RXFPR1 Relaxin receptor 1
Gene Aliases: LGR7; RXFP1; RXFPR1
UniProt ID: (Human) Q9HBX9
Entrez Gene ID: (Human) 59350
Molecular Function: extracellular matrix protein receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.