Invitrogen
Manufacturer Code:PA528759
Catalog # PIPA528759
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to a region within amino acids 693 and 757 of Human GPCR LGR7 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: leucine-rich repeat-containing G protein-coupled receptor 7; Leucine-rich repeat-containing G-protein coupled receptor 7; LGR7 RXFPR1 leucine-rich repeat-containing G protein-coupled receptor 7 relaxin receptor 1; Relaxin family peptide receptor 1; Relaxin receptor 1; relaxin/insulin-like family peptide receptor 1
Gene Aliases: LGR7; RXFP1; RXFPR1
UniProt ID: (Human) Q9HBX9
Entrez Gene ID: (Human) 59350
Molecular Function: extracellular matrix protein receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.