Invitrogen
Manufacturer Code:MA525644
Catalog # PIMA525644
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2A2 |
Immunogen | Human recombit protein fragment corresponding to amino acids 250-550 of LGR5 produced in HEK293T cell |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: FEX GPR49 GPR67 GRP49 HG38 G-protein coupled receptor 49 G-protein coupled receptor 67 G-protein coupled receptor HG38 leucine-rich repeat-containing G protein-coupled receptor 5 leucine-rich repeat-containing G-protein coupled receptor 5 orphan G protein-coupled receptor HG38; follicle expressed hormone; G protein-coupled receptor 49; G-protein coupled receptor 49; G-protein coupled receptor 67; G-protein coupled receptor HG38; leucine-rich repeat containing G protein-coupled receptor 5; leucine-rich repeat-containing G protein-coupled receptor 5; Leucine-rich repeat-containing G-protein coupled receptor 5; orphan G protein-coupled receptor HG38; Orphan G-protein coupled receptor FEX
Gene Aliases: FEX; GPR49; GPR67; GRP49; HG38; LGR5
UniProt ID: (Human) O75473, (Mouse) Q9Z1P4
Entrez Gene ID: (Human) 8549, (Mouse) 14160
Molecular Function: extracellular matrix protein receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.