Invitrogen
Manufacturer Code:PA547244
Catalog # PIPA547244
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombit mouse Legumain Val18-Tyr435 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: AEP Asparaginyl Endopeptidase LGMN AEP Asparaginyl endopeptidase cysteine protease 1 legumain LGMN1 Protease cysteine 1 protease cysteine 1 (legumain) PRSC1EC 3.4.22.34; Asparaginyl endopeptidase; Legumain; preprolegumain; Protease, cysteine 1; protease, cysteine, 1
Gene Aliases: AEP; AI746452; AU022324; Lgmn; Prsc1
UniProt ID: (Mouse) O89017
Entrez Gene ID: (Mouse) 19141
Molecular Function: cysteine protease hydrolase protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.