Invitrogen
Manufacturer Code:PA545866
Catalog # PIPA545866
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human LGMN |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AEP; Asparaginyl endopeptidase; cysteine protease 1; Legumain; legumain AEP LGMN1 PRSC1; preprolegumain; Protease, cysteine 1; protease, cysteine, 1; protease, cysteine, 1 (legumain)
Gene Aliases: AEP; AI746452; AU022324; cb173; cb460; LGMN; LGMN1; PRSC1; sb:cb173; wu:fa98g05; wu:fj92g08; zgc:76953
UniProt ID: (Bovine) Q95M12, (Human) Q99538, (Mouse) O89017, (Rat) Q9R0J8
Entrez Gene ID: (Bovine) 281281, (Rabbit) 100346565, (Dog) 480232, (Human) 5641, (Horse) 100053265, (Mouse) 19141, (Guinea pig) 100718170, (Rat) 63865, (Zebrafish) 406625
Molecular Function: cysteine protease hydrolase protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.