Proteintech
Manufacturer Code:219191AP150UL
Catalog # 50-173-3640
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | LGI3 Fusion Protein Ag16257 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Leubrin; Leucine-rich glioma-inactivated protein 3; Leucine-rich repeat LGI family member 3; leucine-rich repeat LGI family, member 3; LGI1 like protein 4 LGI3 LGIL4; LGI1-like protein 4
Gene Aliases: AI841179; AW049851; LGI3; LGIL4; UNQ8190/PRO23199
UniProt ID: (Human) Q8N145, (Mouse) Q8K406
Entrez Gene ID: (Human) 203190, (Rat) 306013, (Mouse) 213469
Molecular Function:
extracellular matrix protein
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.