Invitrogen
Manufacturer Code:PA542669
Catalog # PIPA542669
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human LFNG |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Beta-1,3-N-acetylglucosaminyltransferase lunatic fringe; LFNG O-fucosylpeptide 3-beta-N-acetylglucosaminyltransferase SCDO3; lunatic fringe; lunatic fringe gene homolog; O-fucosylpeptide 3-beta-N-acetylglucosaminyltransferase
Gene Aliases: AW061165; LFNG; SCDO3
UniProt ID: (Human) Q8NES3, (Bovine) Q2KJ92, (Mouse) O09010, (Rat) Q924T4
Entrez Gene ID: (Human) 3955, (Bovine) 516209, (Mouse) 16848, (Rat) 170905
Molecular Function: glycosyltransferase signaling molecule transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.